1-Bromo-5-chloro-2-iodobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

B25409-06 B25409-14
AAB25409-03EA 43.12 USD
AAB25409-03 AAB25409-06 AAB25409-14
1-Bromo-5-chloro-2-iodobenzene 98%
1-Bromo-5-chloro-2-iodobenzene

Purity: 98% CAS No.: 31928-44-6 Molecular Formula: C6H3BrClI Formula Weight: 317.36 MDL No.: MFCD00672944

Formula: C₆H₃BrClI
MW: 190.45
Storage Temperature: Ambient
MDL Number: MFCD00672944
CAS Number: 31928-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR