5-Fluoro-o-toluidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Amino-4-fluorotoluene, 5-Fluoro-2-methylaniline
B25381-09
B25381-18
AAAB25381-09EA
30.3
USD
AAAB25381-09
AAAB25381-18
5-Fluoro-o-toluidine 98%
5-Fluoro-o-toluidine
Purity: 98%
CAS No.: 367-29-3
Molecular Formula: C7H8FN
Formula Weight: 125.15
Flash Point: 90°(194°F)
Refractive Index: 1.5380
Odor: Characteristic
Melting Point: 35-38°
Boiling Point: 98-100°/15mm
Beilstein Reference: 2637584
UN No.: 2811
MDL No.: MFCD00007764
Formula:
C₇H₈FN MW: 125.15 g/mol Boiling Pt: 98…100 °C/15 mm Melting Pt: 35…38 °C Density: 1.13 g/cm³ (20 °C) Flash Pt: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007764 CAS Number: 367-29-3 EINECS: 206-689-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...