2-Butynoic acid ethyl ester 98%

Supplier: Thermo Scientific Chemicals
Warning

B25370-06 B25370-03 B25370-14
AAB25370-06EA 89.2 USD
AAB25370-06 AAB25370-03 AAB25370-14
2-Butynoic acid ethyl ester 98%
2-Butynoic acid ethyl ester

Purity: 98% CAS No.: 4341-76-8 Molecular Formula: C6H8O2 Formula Weight: 112.13 Flash Point: 53°(127°F) Refractive Index: 1.4380 Odor: Characteristic Boiling Point: 163° Beilstein Reference: 1744948 UN No.: 3272 MDL No.: MFCD00015182

Formula: C₆H₈O₂
Boiling Pt: 163 °C
Density: 0.967
Flash Pt: 58 °C (127 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015182
CAS Number: 4341-76-8
EINECS: 224-395-6
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR