2-Bromo-5-nitroanisole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25331-04
B25331-09
AAAB25331-04EA
33.9
USD
AAAB25331-04
AAAB25331-09
2-Bromo-5-nitroanisole 98%
2-Bromo-5-nitroanisole
Purity: 98%
CAS No.: 77337-82-7
Molecular Formula: C7H6BrNO3
Formula Weight: 232.03
Melting Point: 102-107°
Beilstein Reference: 2520496
Formula:
C₇H₆BrNO₃ Melting Pt: 102…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041250 CAS Number: 77337-82-7 EINECS: 278-669-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...