5-Methoxyindole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25290-03
B25290-06
B25290-14
AAAB25290-03EA
40.9
USD
AAAB25290-03
AAAB25290-06
AAAB25290-14
5-Methoxyindole 99%
5-Methoxyindole
Purity: 99%
CAS No.: 1006-94-6
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Melting Point: 52-55°
Boiling Point: 176-178°/17mm
Beilstein Reference: 116722
MDL No.: MFCD00005674
Formula:
C₉H₉NO Boiling Pt: 176…178 °C/17 mm Melting Pt: 52…55 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005674 CAS Number: 1006-94-6 EINECS: 213-745-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...