(R)-(-)-2-Amino-1-butanol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 98%
CAS No.: 5856-63-3
Molecular Formula: C4H11NO
Formula Weight: 89.14
Flash Point: 89°(192°F)
Refractive Index: 1.4525
Odor: Amine-like
pH: 11.1
Boiling Point: 176-178°
Beilstein Reference: 1718929
Optical Rotation: -10° (neat)
UN No.: 2735
MDL No.: MFCD00064419
Formula:
C₄H₁₁NO Boiling Pt: 176…178 °C Melting Pt: -2 °C Density: 0.936 g/cm³ (20 °C) Flash Pt: 89 °C (192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064419 CAS Number: 5856-63-3 EINECS: 227-476-4 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...