2-Amino-5-iodopyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Iodopyridin-2-amine, 5-Iodo-2-pyridylamine

B25175-06 B25175-14 B25175-03
AAAB25175-06EA 133 USD
AAAB25175-06 AAAB25175-14 AAAB25175-03
2-Amino-5-iodopyridine 97%
2-Amino-5-iodopyridine

Purity: 97% CAS No.: 20511-12-0 Molecular Formula: C5H5IN2 Formula Weight: 220.01 Melting Point: 126-130° Beilstein Reference: 108738

Formula: C₅H₅IN₂
MW: 220.01 g/mol
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00160312
CAS Number: 20511-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR