9-Fluorenone-4-carboxylic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 9-Oxofluoren-4-carboxylic acid, 9-Oxo-9H-fluorene-4-carboxylic acid, 9-Oxofluorene-4-carboxylic acid

B25021-03 B25021-06
AAAB25021-03EA 31.8 USD
AAAB25021-03 AAAB25021-14 AAAB25021-06
9-Fluorenone-4-carboxylic acid 98+%
9-Fluorenone-4-carboxylic acid

Purity: 98+% CAS No.: 6223-83-2 Molecular Formula: C14H8O3 Formula Weight: 224.22 Melting Point: 224-228° Beilstein Reference: 2694801

Formula: C₁₄H₈O₃
MW: 224.22 g/mol
Melting Pt: 224…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00001145
CAS Number: 6223-83-2
EINECS: 228-311-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR