3-Fluorophenyl isothiocyanate 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Fluoro-3-isothiocyanatobenzene
B25001-06
B25001-14
AAAB25001-06EA
56.9
USD
AAAB25001-06
AAAB25001-14
3-Fluorophenyl isothiocyanate 97+%
3-Fluorophenyl isothiocyanate
Purity: 97+%
CAS No.: 404-72-8
Molecular Formula: C7H4FNS
Formula Weight: 153.18
Flash Point: 85°(185°F)
Refractive Index: 1.6200
Odor: Pungent
Boiling Point: 227°
Beilstein Reference: 2081172
UN No.: 2922
MDL No.: MFCD00004804
Formula:
C₇H₄FNS MW: 153.18 g/mol Boiling Pt: 226 - 227 °C (1013 hPa) Density: 1.27 g/cm³ (20 °C) Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004804 CAS Number: 404-72-8 EINECS: 206-967-7 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...