3-Bromobenzylamine 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (3-Bromophenyl)methanamine

B24963-06 B24963-14 B24963-22
AAB24963-06EA 49.9 USD
AAB24963-06 AAB24963-14 AAB24963-22
3-Bromobenzylamine 95%
3-Bromobenzylamine

Purity: 97% CAS No.: 10269-01-9 Molecular Formula: C7H8BrN Formula Weight: 186.06 Refractive Index: 1.5885 Odor: Amine-like UN No.: 2735 MDL No.: MFCD01026119

Formula: C₇H₈BrN
MW: 186.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01026119
CAS Number: 10269-01-9
EINECS: 679-024-4
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR