(±)-1-Pentyn-3-ol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-Ethyl ethynyl carbinol, 1-Pentyn-3-ol

B24853-06 B24853-14
AAAB24853-06EA 53.9 USD
AAAB24853-06 AAAB24853-14
(±)-1-Pentyn-3-ol 98%
(±)-1-Pentyn-3-ol

Purity: 98% CAS No.: 4187-86-4 Molecular Formula: C5H8O Formula Weight: 84.12 Flash Point: 29°(84°F) Refractive Index: 1.4340 Odor: Recognizable Boiling Point: 123-125° Beilstein Reference: 1098409 UN No.: 1986 MDL No.: MFCD00004572

Formula: C₅H₈O
Boiling Pt: 123…125 °C
Density: 0.8926 g/cm³ (15 °C)
Flash Pt: 29 °C (84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004572
CAS Number: 4187-86-4
EINECS: 224-063-0
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR