3,4-Dimethylbenzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Xylene-4-sulphonyl chloride, 3,4-Dimethylbenzene-1-sulphonyl chloride

B24806-06 B24806-14
AAAB24806-06EA 103 USD
AAAB24806-06 AAAB24806-22 AAAB24806-14
3,4-Dimethylbenzenesulfonyl chloride 98%
3,4-Dimethylbenzenesulphonyl chloride

Purity: 98% CAS No.: 2905-30-8 Molecular Formula: C8H9ClO2S Formula Weight: 204.68 Odor: Acrid UN No.: 3261 MDL No.: MFCD00625742

Formula: C₈H₉ClO₂S
MW: 204.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00625742
CAS Number: 2905-30-8
EINECS: 210-337-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR