2,5-Dichlorophenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24787-14
B24787-22
B24787-36
AAAB24787-14EA
34.4
USD
AAAB24787-14
AAAB24787-22
AAAB24787-36
2,5-Dichlorophenol 98%
2,5-Dichlorophenol
Purity: 98%
CAS No.: 583-78-8
Molecular Formula: C6H4Cl2O
Formula Weight: 163.00
Odor: Phenol-like
Melting Point: 55-58°
Boiling Point: 211°
Beilstein Reference: 1907692
UN No.: 2020
MDL No.: MFCD00002174
Formula:
C₆H₄Cl₂O MW: 163 g/mol Boiling Pt: 211 °C Melting Pt: 55…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002174 CAS Number: 583-78-8 EINECS: 209-520-4 UN: 2020 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...