1-Methylcyclopentanol 98%

Supplier: Thermo Scientific Chemicals
Warning

B24744-22 B24744-06
AAAB24744-22EA 761 USD
AAAB24744-22 AAAB24744-06 AAAB24744-14
1-Methylcyclopentanol 98%
1-Methylcyclopentanol

Purity: 98% CAS No.: 1462-03-9 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: 40°(105°F) Melting Point: 36-37° Boiling Point: 135-136° UN No.: 1325 MDL No.: MFCD00001364

Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 135 °C (1013 hPa)
Melting Pt: 32 - 33 °C
Density: 0.905 g/cm³ (23.5 °C)
Flash Pt: 40 °C (105 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001364
CAS Number: 1462-03-9
EINECS: 215-963-4
UN: 1987
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR