3,4-Dichlorobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Danger

B24706-06 B24706-14
AAAB24706-06EA 24.2 USD
AAAB24706-06 AAAB24706-14
3,4-Dichlorobenzaldehyde 97%
3,4-Dichlorobenzaldehyde

Purity: 97% CAS No.: 6287-38-3 Molecular Formula: C7H4Cl2O Formula Weight: 175.01 Flash Point: 135°(275°F) Melting Point: 41-44° Boiling Point: 247-248° Beilstein Reference: 636379 UN No.: 1759 MDL No.: MFCD00003351

Formula: C₇H₄Cl₂O
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 43 °C
Flash Pt: 135 °C (275 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003351
CAS Number: 6287-38-3
EINECS: 228-520-5
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR