Ethyl toluene-4-sulfonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 4-toluenesulphonate, Ethyl p-toluenesulphonate, p-Toluenesulphonic acid ethyl ester, Ethyl 4-methylbenzenesulphonate, Toluene-4-sulphonic acid ethyl ester

B24658-18 B24658-30 B24658-0B
AAB24658-18EA 27.8 USD
AAB24658-18 AAB24658-30 AAB24658-0B
Ethyl toluene-4-sulfonate 98%
Ethyl toluene-4-sulphonate

Purity: 98% CAS No.: 80-40-0 Molecular Formula: C9H12O3S Formula Weight: 200.26 Flash Point: 157°(314°F) Refractive Index: 1.5110 Melting Point: 29-34° Boiling Point: 221° Beilstein Reference: 611213 Merck Reference: 14,3858 UN No.: 2811 MDL No.: MFCD00009100

Formula: C₉H₁₂O₃S
MW: 200.26 g/mol
Boiling Pt: 221 °C
Melting Pt: 29…34 °C
Density: 1.174 g/cm³ (20 °C)
Flash Pt: 157 °C(314 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009100
CAS Number: 80-40-0
EINECS: 201-276-7
ADR: 6.1,III
Merck Index: 13,03889

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR