2-Aminopyrimidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pyrimidin-2-ylamine, Pyrimidin-2-amine
B24594-22
B24594-36
AAB24594-22EA
39.1
USD
AAB24594-22
AAB24594-36
2-Aminopyrimidine 98%
2-Aminopyrimidine
Purity: 98%
CAS No.: 109-12-6
Molecular Formula: C4H5N3
Formula Weight: 95.11
Melting Point: 123-127°
Boiling Point: 159°/186mm
MDL No.: MFCD00006089
Formula:
C₄H₅N₃ MW: 95.1 g/mol Boiling Pt: 159 °C/186 mm Melting Pt: 123…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006089 CAS Number: 109-12-6 EINECS: 203-648-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...