3-Methoxyphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-Methoxyphenyl)metaboric acid, Dihydroxy(3-methoxyphenyl)borane, 3-Methoxybenzeneboronic acid

B24412-14 B24412-03 B24412-06
AAB24412-14EA 446 USD
AAB24412-14 AAB24412-03 AAB24412-06
3-Methoxyphenylboronic acid 97%
3-Methoxyphenylboronic acid

Purity: 97% CAS No.: 10365-98-7 Molecular Formula: C7H9BO3 Formula Weight: 151.96 Melting Point: 158-162° Beilstein Reference: 2831223

Formula: C₇H₉BO₃
MW: 151.96 g/mol
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00161359
CAS Number: 10365-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR