Methyl dichloroacetate 99%

Supplier: Thermo Scientific Chemicals
Warning

B24399-22 B24399-36
AAB24399-22EA 38.55 USD
AAB24399-22 AAB24399-36
Methyl dichloroacetate 99%
Methyl dichloroacetate

Purity: 99% CAS No.: 116-54-1 Molecular Formula: C3H4Cl2O2 Formula Weight: 142.97 Flash Point: 70°(158°F) Refractive Index: 1.4420 Odor: Pungent Melting Point: -52° Boiling Point: 142-144° UN No.: 2299 MDL No.: MFCD00000843

Formula: C₃H₄Cl₂O₂
MW: 142.97 g/mol
Boiling Pt: 142…144 °C
Melting Pt: –52 °C
Density: 1.3774 g/cm³ (20 °C)
Flash Pt: 70 °C (158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000843
CAS Number: 116-54-1
EINECS: 204-146-8
UN: 2299
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR