(±)-2-Aminobutan-1-ol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Aminobutan-1-ol, (±)-2-Amino-1-butanol
B24398-AP
B24398-AE
AAB24398-APEA
175
USD
AAB24398-AP
AAB24398-AE
(±)-2-Aminobutan-1-ol 97%
(±)-2-Aminobutan-1-ol
Purity: 97%
CAS No.: 96-20-8
Molecular Formula: C4H11NO
Formula Weight: 89.14
Flash Point: 89°(192°F)
Refractive Index: 1.4525
Odor: Amine-like
pH: 11.1
Melting Point: -2°
Boiling Point: 176-178°
Beilstein Reference: 1098274
Merck Reference: 14,427
UN No.: 2735
MDL No.: MFCD00008095
Formula:
C₄H₁₁NO MW: 89.14 g/mol Boiling Pt: 176 °C (1013 hPa) Melting Pt: -2 °C Density: 0.944 g/cm³ (20 °C) Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008095 CAS Number: 96-20-8 EINECS: 202-488-2 UN: 2735 ADR: 8,III Merck Index: 13,00426 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...