2-Chloro-5-nitrobenzonitrile 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24287-14
B24287-06
B24287-22
AAB24287-14EA
101
USD
AAB24287-14
AAB24287-06
AAB24287-22
2-Chloro-5-nitrobenzonitrile 99%
2-Chloro-5-nitrobenzonitrile
Purity: 99%
CAS No.: 16588-02-6
Molecular Formula: C7H3ClN2O2
Formula Weight: 182.57
Melting Point: 104-107°
Beilstein Reference: 1818642
Formula:
C₇H₃ClN₂O₂ Boiling Pt: 119 - 122 °C (0.6 -rr) Melting Pt: 104…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007292 CAS Number: 16588-02-6 EINECS: 240-643-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...