1,5-Diaminonaphthalene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,5-naphthylenediamine, 1,5-Naphthalenediamine

B24262-14 B24262-22
AAB24262-14EA 57.6 USD
AAB24262-14 AAB24262-22
1,5-Diaminonaphthalene 97%
1,5-Diaminonaphthalene

Purity: 97% CAS No.: 2243-62-1 Molecular Formula: C10H10N2 Formula Weight: 158.20 Melting Point: 189-192° Boiling Point: 200-210°/5mm UN No.: 3077 MDL No.: MFCD00004029

Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 200…210 °C/5 mm
Melting Pt: 189…192 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004029
CAS Number: 2243-62-1
EINECS: 218-817-8
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR