4'-Bromo-4-chlorobutyrophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24146-22
B24146-14
AAB24146-22EA
145
USD
AAB24146-22
AAB24146-14
4'-Bromo-4-chlorobutyrophenone 98%
4'-Bromo-4-chlorobutyrophenone
Purity: 98%
CAS No.: 4559-96-0
Molecular Formula: C10H10BrClO
Formula Weight: 261.55
Flash Point:
Formula:
C₁₀H₁₀BrClO Boiling Pt: 135 - 139 °C (0.3 -rr) Melting Pt: 35…38 °C Density: 1.4808 g/cm³ (20 °C) Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001006 CAS Number: 4559-96-0 EINECS: 224-930-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...