N,N-Dimethyldodecylamine 90+%

Supplier: Thermo Scientific Chemicals
Danger

B24145-0B B24145-18 B24145-30
AAB24145-0BEA 262 USD
AAB24145-0B AAB24145-18 AAB24145-30
N,N-Dimethyldodecylamine 90+%
N,N-Dimethyldodecylamine

Purity: 90+% CAS No.: 112-18-5 Molecular Formula: CH3(CH2)11N(CH3)2 Formula Weight: 213.41 Flash Point: 118°(244°F) Refractive Index: 1.4375 Melting Point: -20° Boiling Point: 110-112°/3mm UN No.: 2735 MDL No.: MFCD00008970

Formula: C₁₄H₃₁N
Boiling Pt: 110…112 °C/3 mm
Melting Pt: –20 °C
Density: 0.787 g/cm³ (20 °C)
Flash Pt: 118 °C (244 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008970
CAS Number: 112-18-5
EINECS: 203-943-8
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR