5-Amino-1H-tetrazole monohydrate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Aminotetrazole monohydrate, 5-Amino-1H-tetrazole monohydrate, Tetrazol-5-amine, 1H-Tetrazol-5-ylamine monohydrate, 1H-Tetrazol-5-amine hydrate, 5-Amino-tetrazole-monohydrate

B24018-22 B24018-36 B24018-14
AAB24018-22EA 65.9 USD
AAB24018-22 AAB24018-36 AAB24018-14
5-Amino-1H-tetrazole monohydrate 99%
5-Amino-1H-tetrazole monohydrate

Purity: 99% CAS No.: 15454-54-3 Molecular Formula: CH3N5.H2O Formula Weight: 103.08 (85.06anhy) pH: 3.7 Melting Point: ca 203° dec. Beilstein Reference: 108872 UN No.: 1325 MDL No.: MFCD00149327

Formula: CH₃N₅·1H₂O
MW: 103.08 g/mol
Melting Pt: 216 - 217 °C
Density: 1.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149327
CAS Number: 15454-54-3
EINECS: 224-581-7
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR