Benzenesulphonohydrazide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Benzenesulphonic acid hydrazide, Benzenesulphonyl hydrazide

B23971-36 B23971-14 B23971-22
AAB23971-36EA 531 USD
AAB23971-36 AAB23971-14 AAB23971-22
Benzenesulphonohydrazide 98%
Benzenesulphonohydrazide

Purity: 98% CAS No.: 80-17-1 Molecular Formula: C6H8N2O2S Formula Weight: 172.21 Melting Point: 101-105° Beilstein Reference: 640079 Merck Reference: 14,7598 UN No.: 3226 MDL No.: MFCD00007583

Formula: C₆H₈N₂O₂S
Melting Pt: 101…110 °C (dec.)
Density: 1.369 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007583
CAS Number: 80-17-1
EINECS: 201-255-2
UN: 3226
ADR: 4.1,
Merck Index: 12,07757

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR