Isophthalonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Benzenedicarbonitrile, 1,3-Dicyanobenzene, Benzene-1,3-dicarbonitrile

B23900-30 B23900-0B
AAB23900-30EA 46.7 USD
AAB23900-30 AAB23900-0B
Isophthalonitrile 98%
Isophthalonitrile

Purity: 98% CAS No.: 626-17-5 Molecular Formula: C8H4N2 Formula Weight: 128.13 Flash Point: 150°(302°F) Odor: Almond-like Boiling Point: 288° Beilstein Reference: 1072209 MDL No.: MFCD00001795

Formula: C₈H₄N₂
MW: 128.13 g/mol
Boiling Pt: 288 °C
Melting Pt: 159…163 °C
Density: 4.42 g/cm³ (20 °C)
Flash Pt: >150 °C (302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001795
CAS Number: 626-17-5
EINECS: 210-933-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR