2-Phenylimidazole 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Phenyl-1H-imidazole

B23858-14 B23858-22
AAAB23858-14EA 44.6 USD
AAAB23858-14 AAAB23858-22
2-Phenylimidazole 98+%
2-Phenylimidazole

Purity: 98+% CAS No.: 670-96-2 Molecular Formula: C9H8N2 Formula Weight: 144.18 Flash Point: 200°(392°F) Melting Point: 146-150° Boiling Point: 198-200°/7mm Beilstein Reference: 114011 MDL No.: MFCD00005186

Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 143 - 146 °C
Density: 0.6 g/cm³ (20 °C)
Flash Pt: 200 °C (392 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005186
CAS Number: 670-96-2
EINECS: 211-581-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR