2'-Chloro-5'-(trifluoromethyl)acetophenone 97+%

Supplier: Thermo Scientific Chemicals
Warning

B23524-06 B23524-03
AAAB23524-06EA 115 USD
AAAB23524-06 AAAB23524-03
2'-Chloro-5'-(trifluoromethyl)acetophenone 97+%
2'-Chloro-5'-(trifluoromethyl)acetophenone

Purity: 97+% CAS No.: 71648-45-8 Molecular Formula: C9H6ClF3O Formula Weight: 222.60 MDL No.: MFCD00045092

Formula: C₉H₆ClF₃O
MW: 222.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045092
CAS Number: 71648-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR