(±)-Mercaptosuccinic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23301-36
B23301-14
B23301-22
AAAB23301-36EA
159
USD
AAAB23301-36
AAB23301-14
AAB23301-22
(±)-Mercaptosuccinic acid 98%
(±)-Mercaptosuccinic acid
Purity: 98%
CAS No.: 70-49-5
Molecular Formula: C4H6O4S
Formula Weight: 150.15
Melting Point: 150-155°
Beilstein Reference: 1099858
Merck Reference: 14,9343
MDL No.: MFCD00004860
Formula:
C₄H₆O₄S Boiling Pt: 322 °C (1013 hPa) Melting Pt: 150 - 152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004860 CAS Number: 70-49-5 EINECS: 200-736-4 Merck Index: 12,09479 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...