(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, (2S)-(-)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid
B23117-ME
B23117-88
AAAB23117-MEEA
97.8
USD
AAAB23117-ME
AAAB23117-88
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid
Purity: 99%
CAS No.: 17257-71-5
Molecular Formula: C10H9F3O3
Formula Weight: 234.18
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₁₀H₉F₃O₃ MW: 234.17 g/mol Boiling Pt: 105…107 °C/1 mm Melting Pt: 43…45 °C Density: 1.344 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064200 CAS Number: 17257-71-5 EINECS: 241-292-1 Merck Index: 14,6280 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...