2,3-Dichlorophenol 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22943-22
B22943-14
B22943-36
AAB22943-22EA
80.4
USD
AAB22943-22
AAB22943-14
AAB22943-36
2,3-Dichlorophenol 98+%
2,3-Dichlorophenol
Purity: 98+%
CAS No.: 576-24-9
Molecular Formula: C6H4Cl2O
Formula Weight: 163.00
Flash Point: 115°(239°F)
Odor: Phenol-like
Boiling Point: 206°
Beilstein Reference: 2043615
UN No.: 2020
MDL No.: MFCD00002160
Formula:
C₆H₄Cl₂O MW: 163 g/mol Boiling Pt: 206 °C Melting Pt: 57…60 °C Density: 1.4 g/cm³ (25 °C) Flash Pt: 115 °C (239 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002160 CAS Number: 576-24-9 EINECS: 209-399-8 UN: 2928 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...