3-Bromo-1-propanol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Bromopropan-1-ol, 1-Bromo-3-hydroxypropane
B22857-22
B22857-06
B22857-14
AAAB22857-22EA
338
USD
AAAB22857-22
AAAB22857-06
AAAB22857-14
3-Bromo-1-propanol 97%
3-Bromo-1-propanol
Purity: 97+%
CAS No.: 627-18-9
Molecular Formula: C3H7BrO
Formula Weight: 138.99
Storage Temperature: 2°C to 8°C
Flash Point: 93°(199°F)
Refractive Index: 1.4885
Boiling Point: 71-73°/11mm
Beilstein Reference: 969160
MDL No.: MFCD00002942
Formula:
BrH₂CCH₂CH₂OH MW: 138.99 g/mol Boiling Pt: 71…73 °C/11 mm Density: 1.6 g/cm³ (20 °C) Flash Pt: 93 °C(199 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002942 CAS Number: 627-18-9 EINECS: 210-986-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...