1-Chloro-3,4-dinitrobenzene tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 90%
CAS No.: 610-40-2
Molecular Formula: C6H3ClN2O4
Formula Weight: 202.55
Flash Point:
Formula:
C₆H₃ClN₂O₄ Boiling Pt: 14…16 °C/4 mm Melting Pt: ∼ 20…25 °C Density: 1.687 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007212 CAS Number: 610-40-2 EINECS: 210-223-7 UN: 1577 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...