Ethyl-2-ethylacetoacetate 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethyl 2-ethyl-3-oxobutanoate
B22295-14
B22295-22
AAB22295-14EA
107
USD
AAB22295-14
AAB22295-22
Ethyl-2-ethylacetoacetate 95%
Ethyl-2-ethylacetoacetate
Purity: 95%
CAS No.: 607-97-6
Molecular Formula: C8H14O3
Formula Weight: 158.20
Flash Point: 77°(170°F)
Refractive Index: 1.4220
Boiling Point: 83-85°/10mm
Beilstein Reference: 636286
MDL No.: MFCD00039898
Formula:
C₈H₁₄O₃ MW: 158.2 Boiling Pt: 83…85 °C/10 mm Density: 0.9937 Flash Pt: 77 °C(170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039898 CAS Number: 607-97-6 EINECS: 210-151-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...