Allylchlorodimethylsilane 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Allyldimethylchlorosilane, ADMCS

B22237-09 B22237-06 B22237-14
AAAB22237-09EA 137 USD
AAAB22237-09 AAB22237-06 AAB22237-14
Allylchlorodimethylsilane 95%
Allylchlorodimethylsilane

Purity: 95% CAS No.: 4028-23-3 Molecular Formula: CH2=CHCH2Si(CH3)2Cl Formula Weight: 134.68 Flash Point: 5°(41°F) Refractive Index: 1.4290 Boiling Point: 110-112° Beilstein Reference: 1737978 UN No.: 2985 MDL No.: MFCD00000506

Formula: C₅H₁₁ClSi
MW: 134.68 g/mol
Boiling Pt: 110…112 °C
Density: 0.896
Flash Pt: 5 °C(41 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000506
CAS Number: 4028-23-3
EINECS: 223-706-2
UN: 2985
ADR: "3",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR