Sodium triacetoxyborohydride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22060-14
B22060-22
B22060-06
AAAB22060-14EA
95.37
USD
AAAB22060-14
AAAB22060-22
AAAB22060-06
Sodium triacetoxyborohydride 95%
Sodium triacetoxyborohydride
Purity: 95%
CAS No.: 56553-60-7
Molecular Formula: Na(CH3CO2)3BH
Formula Weight: 211.94
Melting Point: ca 110° dec.
Beilstein Reference: 4047608
UN No.: 3396
MDL No.: MFCD00012211
Formula:
C₆H₁₀BNaO₆ MW: 211.94 g/mol Melting Pt: ca 110 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012211 CAS Number: 56553-60-7 UN: 3396 ADR: "4.3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...