Cobalt(II) hydroxide 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cobalt dihydroxide, Cobaltous hydroxide, Cobalt(II) hydroxide

B22045-18 B22045-30
AAB22045-18EA 44.89 USD
AAB22045-18 AAB22045-30
Cobalt(II) hydroxide 97%
Cobalt(II) hydroxide

Purity: 97% CAS No.: 21041-93-0 Molecular Formula: H2CoO2 Formula Weight: 92.95 Odor: Odorless Melting Point: 250° dec. MDL No.: MFCD00016015

Formula: H₂CoO₂
MW: 92.95
Melting Pt: 250 °C (dec.)
Density: 3.597
Storage Temperature: Ambient
MDL Number: MFCD00016015
CAS Number: 21041-93-0
EINECS: 244-166-4
UN: 3077
ADR: 9 III
Merck Index: 14,2442

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR