2-Amino-3,5-dichloropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,5-Dichloro-2-pyridylamine, 3,5-Dichloropyridin-2-amine
B21984-22
B21984-14
AAAB21984-06EA
33.19
USD
AAAB21984-06
AAAB21984-22
AAAB21984-14
2-Amino-3,5-dichloropyridine 98%
2-Amino-3,5-dichloropyridine
Purity: 98%
CAS No.: 4214-74-8
Molecular Formula: C5H4Cl2N2
Formula Weight: 163.01
Odor: Unpleasant
Melting Point: 81-83°
Beilstein Reference: 119376
MDL No.: MFCD00006313
Formula:
C₅H₄Cl₂N₂ Melting Pt: 81…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006313 CAS Number: 4214-74-8 EINECS: 224-143-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...