4-Bromo-2,6-dimethylanisole 99%

Supplier: Thermo Scientific Chemicals
Warning

B21931-14 B21931-06
AAB21931-14EA 47.1 USD
AAB21931-14 AAB21931-06 AAB21931-22
4-Bromo-2,6-dimethylanisole 99%
4-Bromo-2,6-dimethylanisole

Purity: 99% CAS No.: 14804-38-7 Molecular Formula: C9H11BrO Formula Weight: 215.09 Flash Point: 110°(230°F) Refractive Index: 1.5450 Boiling Point: 117-120°/13mm Beilstein Reference: 1940290 MDL No.: MFCD00143259

Formula: C₉H₁₁BrO
MW: 215.09 g/mol
Boiling Pt: 117 - 120 °C (13 -rr)
Density: 1.347
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00143259
CAS Number: 14804-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR