3-Hydroxybenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: m-Hydroxybenzonitrile, m-Cyanophenol, 3-Cyanophenol

B21702-06 B21702-14
AAB21702-06EA 41.1 USD
AAB21702-06 AAB21702-14
3-Hydroxybenzonitrile 99%
3-Hydroxybenzonitrile

Purity: 99% CAS No.: 873-62-1 Molecular Formula: C7H5NO Formula Weight: 119.12 Melting Point: 78-81° MDL No.: MFCD00002252

Formula: C₇H₅NO
MW: 119.12 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 84.5 - 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00002252
CAS Number: 873-62-1
EINECS: 212-845-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR