Trichloroacetyl chloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21675-0B
B21675-18
B21675-30
AAB21675-0BEA
909
USD
AAB21675-0B
AAB21675-18
AAB21675-30
Trichloroacetyl chloride 99%
Trichloroacetyl chloride
Purity: 99%
CAS No.: 76-02-8
Molecular Formula: CCl3COCl
Formula Weight: 181.83
Refractive Index: 1.4700
Melting Point: -146°
Boiling Point: 114-116°
Beilstein Reference: 774120
UN No.: 2442
MDL No.: MFCD00000792
Formula:
C₂Cl₄O MW: 181.83 Boiling Pt: 118 Melting Pt: -57 Density: 1.629 Storage Temperature: Ambient |
MDL Number:
MFCD00000792 CAS Number: 76-02-8 EINECS: 200-926-7 UN: 2442 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...