Cesium formate 98%

Supplier: Thermo Scientific Chemicals
Warning

B21327-14 B21327-22
AAAB21327-14EA 57.16 USD
AAAB21327-14 AAAB21327-22
Cesium formate 98%
Cesium formate

Purity: 98% CAS No.: 3495-36-1 Molecular Formula: CsOOCH Formula Weight: 177.92 Beilstein Reference: 3912426 MDL No.: MFCD00039103

Formula: CHCsO₂
Storage Temperature: Ambient
MDL Number: MFCD00039103
CAS Number: 3495-36-1
EINECS: 222-492-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR