Chlorobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzene ChloridePhenylchlorideMonochlorobenzene

B21052-0F B21052-AP
AAAB21052-APEA 51.15 USD
AAAB21052-AP AAAB21052-0F
Chlorobenzene 99%
Chlorobenzene
Formula: C₆H₅Cl
MW: 112.56 g/mol
Boiling Pt: 132 °C
Melting Pt: –45 °C
Density: 1.106
Flash Pt: 28 °C(82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: "3",III
REACH: 01-2119432722-45
Merck Index: 14,2121
https://pr.vwr.com/stibo/web/std.lang.all/08/56/750856.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR