2,4-Quinolinediol 97%

Supplier: Thermo Scientific Chemicals
Warning

B20747-06 B20747-14
AAAB20747-06EA 31.8 USD
AAAB20747-06 AAAB20747-14
2,4-Quinolinediol 97%
2,4-Quinolinediol

MDL: MFCD00006744 Beilstein: 129767 Insoluble in water


Purity: 97% CAS No.: 86-95-3 Molecular Formula: C9H7NO2 Formula Weight: 161.16 Melting Point: 300° Solubility: Insoluble in water Beilstein Reference: 129767 MDL No.: MFCD00006744

Formula: C₉H₇NO₂
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 159 °C
Storage Temperature: Ambient
MDL Number: MFCD00006744
CAS Number: 86-95-3
EINECS: 201-711-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR