4-Ethylbenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
p-Ethylbenzaldehyde
B20645-22
B20645-14
AAAB20645-22EA
148
USD
AAAB20645-22
AAAB20645-14
4-Ethylbenzaldehyde 97%
4-Ethylbenzaldehyde
Purity: 97%
CAS No.: 4748-78-1
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 92°(197°F)
Refractive Index: 1.5390
Odor: Characteristic
Boiling Point: 220-221°
Beilstein Reference: 636027
MDL No.: MFCD00006956
Formula:
C₉H₁₀O MW: 134.18 g/mol Boiling Pt: 220…221 °C Density: 0.97 g/cm³ (20 °C) Flash Pt: 92 °C(197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006956 CAS Number: 4748-78-1 EINECS: 225-268-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...