Palmitic acid 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Hexadecanoic acid, Hexadecylic acid

B20322-36 B20322-0E B20322-0C
AAAB20322-36EA 66.53 USD
AAAB20322-36 AAAB20322-0E AAAB20322-0C
Palmitic acid 95%
Palmitic acid

A saturated fatty acid


Purity: 95% CAS No.: 57-10-3 Molecular Formula: C16H32O2 Formula Weight: 256.43 Flash Point: 206°(402°F) Refractive Index: 1.4273 Odor: Odorless Melting Point: 62-65° Boiling Point: 215°/15mm Beilstein Reference: 607489 Merck Reference: 14,6996 MDL No.: MFCD00002747

Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 61 - 63 °C
Density: 0.853 g/cm³ (20 °C)
Flash Pt: 206 °C(402 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002747
CAS Number: 57-10-3
EINECS: 200-312-9
Merck Index: 13,07064

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR