6-Fluoro-m-cresol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Fluoro-5-methylphenol, 1-Fluoro-2-hydroxy-4-methylbenzene

B20275-03 B20275-06
AAAB20275-03EA 31 USD
AAAB20275-03 AAAB20275-06
6-Fluoro-m-cresol 97%
6-Fluoro-m-cresol

Purity: 97% CAS No.: 63762-79-8 Molecular Formula: C7H7FO Formula Weight: 126.13 Flash Point: 67°(152°F) Refractive Index: 1.5105 Boiling Point: 172-174° Beilstein Reference: 2433017 UN No.: 3265 MDL No.: MFCD00190101

Formula: C₇H₇FO
Boiling Pt: 172…174 °C
Density: 1.155
Flash Pt: 67 °C (152 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190101
CAS Number: 63762-79-8
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR