Potassium titanate

Supplier: Thermo Scientific Chemicals

B20263-22 B20263-36
AAB20263-0EEA 677.67 USD
AAB20263-0E AAB20263-22 AAB20263-36
Potassium titanate
Potassium titanate

CAS No.: 12030-97-6 Molecular Formula: K2O + TiO2 Odor: Odorless Melting Point: 1615° MDL No.: MFCD00058803

Formula: K₂O₃Ti
MW: 174.07
Melting Pt: 826
Density: 3.1
Storage Temperature: Ambient
MDL Number: MFCD00058803
CAS Number: 12030-97-6
EINECS: 234-748-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR