2-Chloro-4-nitrobenzamide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Aklomide
A19702-14
A19702-06
AAAA19702-14EA
226
USD
AAAA19702-14
AAAA19702-06
2-Chloro-4-nitrobenzamide 98%
2-Chloro-4-nitrobenzamide
Purity: 98%
CAS No.: 3011-89-0
Molecular Formula: C7H5ClN2O3
Formula Weight: 200.58
Melting Point: 168-172°
Beilstein Reference: 1966537
Formula:
C₇H₅ClN₂O₃ Melting Pt: 168…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017119 CAS Number: 3011-89-0 EINECS: 221-143-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...